Orders LinkContactUsLinkComponent
Replacement mini union ferrules, SGE
Replacement mini union ferrules, SGE
Replacement mini union ferrule, 15% graphite 85% vespel®, 0,3 mm hole
Catalog # 568-0479
Replacement mini union ferrules, SGE
Catalog # 568-0479
Replacement mini union ferrules, SGE
Replacement mini union ferrule, 15% graphite 85% vespel®, 0,3 mm hole
Price
97,90 €
Log in for availability
1 * 10 ST
Product Details & Documents
Specifications
  • Description:
    Replacement mini union ferrule, 15% graphite 85% vespel®, 0,3 mm hole