Orders LinkContactUsLinkComponent
Replacement mini union ferrules, SGE
Replacement mini union ferrules, SGE
Replacement mini union ferrule, 100% graphite, for 0,1 - 0,32 mm ID columns
Catalog # 558-0189
Replacement mini union ferrules, SGE
Catalog # 558-0189
Replacement mini union ferrules, SGE
Replacement mini union ferrule, 100% graphite, for 0,1 - 0,32 mm ID columns
Price
59,30 €
Log in for availability
1 * 10 ST
Product Details & Documents
Specifications
  • Description:
    Replacement mini union ferrule, 100% graphite, for 0,1 - 0,32 mm ID columns