Replacement mini union ferrules, SGE
Replacement mini union ferrule, 100% graphite, for 0,45 - 0,53 mm ID columns
Catalog # 558-0188
Supplier: Trajan Scientific and Medical
Jump to:
Product Details & Documents
Specifications
- Description:Replacement mini union ferrule, 100% graphite, for 0,45 - 0,53 mm ID columns