Order Entry
Belgium
Orders LinkContactUsLinkComponent
Selenium, Supelco®

Selenium, Supelco®

Plastic bottle, 50 g
Catalog # 1.07714.0050
Toxic
Health
Supplier:   Merck
Selenium, Supelco®
Catalog # 1.07714.0050
Supplier:  Merck
CAS Number:  7782-49-2
Selenium, Supelco®

Plastic bottle, 50 g

CAS Number:  7782-49-2
Price
162,00 €
Log in for availability
1 * 50 g
Product Details & Documents
  • CAS Number:7782-49-2
  • Formula:Se
  • MDL Number:MFCD00134090
  • Molecular Weight:78.96 g/mol
  • Merck Index:12,08572
  • Boiling Pt:∼685 °C (1013 hPa)
  • Melting Pt:217 °C
  • Density:4.28 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-957-4
  • ADR:6.1,III
  •  :M-Clarity™ quality level = MQ300
  • UN:3288

Documents

Certificates
Search by Lot Number
Specifications

Specification Test Results

  • Solubility:
    (20 °C) slightly soluble
  • Melting Point:
    217 °C
  • Molar Mass:
    78.96 g/mol
  • Bulk Density:
    1270 kg/m3
  • Boiling Point:
    685 °C (1013 hPa)
  • Vapor Pressure:
    0.001 hPa (20 °C)

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.