ContactUsLinkComponent
SGE SYRINGE 100µL REMOVABLE NEEDLE NEEDLE IS STAINLESS STEEL 50 MM LONG 25 GAUGE (0.50 MM OD) WITH A BEVEL POINT STYLE. BARREL HAS A PTFE SEALING GLAND AT THE REAR FLANGE
Catalog # SGEA005610
SGE SYRINGE 100µL REMOVABLE NEEDLE NEEDLE IS STAINLESS STEEL 50 MM LONG 25 GAUGE (0.50 MM OD) WITH A BEVEL POINT STYLE. BARREL HAS A PTFE SEALING GLAND AT THE REAR FLANGE
Catalog # SGEA005610
Frequently Bought Together