Order Entry
Belgium
Orders LinkContactUsLinkComponent
Syringes for CTC analytics systems

Syringes for CTC analytics systems

Removable needle, 26, 100 µl, 50 mm, 0,47 mm, Conical, 100R-C/T-GT-5/0.47C *
Catalog # 548-1632
Syringes for CTC analytics systems
Catalog # 548-1632
Syringes for CTC analytics systems
Syringes for CTC analytics systems

Removable needle, 26, 100 µl, 50 mm, 0,47 mm, Conical, 100R-C/T-GT-5/0.47C *

Price
138,00 €
Log in for availability
1 * 1 ST
Specifications
  • Ext.Ø:
    0,47 mm
  • Gauge:
    26
  • Type:
    100R-C/T-GT-5/0.47C *
  • Type of tip:
    Conical
  • Volume:
    100 µl
  • Description:
    Removable needle
  • Length:
    50 mm
  • Seq:
    43
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.