ContactUsLinkComponent
SGE SYRINGE 10µL REMOVABLE NEEDLE FOR AGILENT 7673/7683 AUTOSAMPLER NEEDLE IS STAINLESS STEEL 0.63 MM OD (23 GAUGE) 42 MM LONG W/ CO NE POINT STYLE. PLUNGER IS MADE OF A NICKEL/TITANIUM ALLOY THAT WILL NOT PERMANENTLY BEND
Catalog # 549-0396
SGE SYRINGE 10µL REMOVABLE NEEDLE FOR AGILENT 7673/7683 AUTOSAMPLER NEEDLE IS STAINLESS STEEL 0.63 MM OD (23 GAUGE) 42 MM LONG W/ CO NE POINT STYLE. PLUNGER IS MADE OF A NICKEL/TITANIUM ALLOY THAT WILL NOT PERMANENTLY BEND
Catalog # 549-0396
Frequently Bought Together