SGE SYRINGE 10µL REMOVABLE NEEDLE FOR AGILENT 7673/7683 AUTOSAMPLER GAS TIGHT PLUNGER NEEDLE IS STAINLESS STEEL 0.63 MM OD (23 GAUGE ) 42 MM LONG W/ CONE POINT STYLE
Catalog # 549-0386
Supplier: Trajan Scientific and Medical
Frequently Bought Together