Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Syringes for CTC analytics systems

Fixed needle, 26, 5 µl, 50 mm, 0,47 mm, Conical, 5F-C/T-5/0.47C
  548-1583
Syringes for CTC analytics systems
  548-1583
Syringes for CTC analytics systems
Syringes for CTC analytics systems

Fixed needle, 26, 5 µl, 50 mm, 0,47 mm, Conical, 5F-C/T-5/0.47C

 
124,00 €
   
1 * 1 ST
 
  • Ext.Ø:
    0,47 mm
  • Gauge:
    26
  • Type:
    5F-C/T-5/0.47C
  • Type of tip:
    Conical
  • Volume:
    5 µl
  • Description:
    Fixed needle
  • Length:
    50 mm
  • Seq:
    19
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.