Order Entry
Belgium
Orders LinkContactUsLinkComponent
Uridine

Uridine

Glass bottle for solids, 100 g
Catalog # SERA37850.03
Marketsource
Supplier:   Serva
Uridine
Catalog # SERA37850.03
Supplier:  Serva
Marketsource
CAS Number:  58-96-8
Uridine
Uridine

Glass bottle for solids, 100 g

CAS Number:  58-96-8
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside
  • CAS Number:58-96-8
  • Formula:C₉H₁₂N₂O₆
  • MDL Number:MFCD00006526
  • Molecular Weight:244.20 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-407-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications