Order Entry
Belgium
Orders LinkContactUsLinkComponent
Uridine

Uridine

5 g
Catalog # SERA37850.01
Uridine
Catalog # SERA37850.01
Supplier:  
CAS Number:  58-96-8
Uridine
CAS Number:  58-96-8
Discontinued
Product Details & Documents
Synonyms:  1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside
  • CAS Number:58-96-8
  • Formula:C₉H₁₂N₂O₆
  • MDL Number:MFCD00006526
  • Molecular Weight:244.20 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-407-5
Specifications

Specifications

  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.