3,3',5,5'-Tetramethylbenzidine (TMB), research grade
Glass bottle for solids, 5 g
Catalog # SERA35926.02
Marketsource
Supplier: Serva
3,3',5,5'-Tetramethylbenzidine (TMB), research grade
Catalog # SERA35926.02
Supplier: Serva
Marketsource
CAS Number: 54827-17-7
Jump to:
Product Details & Documents
Substrate for horseradish peroxidase.
More sensitive than ABTS and more stable and less toxic than diaminobenzidine. See also TMB Ready-To-Use ELISA Substrate (cat. no. 37068) and SERVAColor TMB Blot Solution (cat. no. 37071).
Synonyms: TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
- CAS Number:54827-17-7
- Formula:C₁₆H₂₀N₂
- MDL Number:MFCD00007748
- Molecular Weight:240.35 g/mol
- Boiling Pt:402 °C (1013 hPa)
- Melting Pt:166…170 °C
- Storage Temperature:Ambient
- EINECS:259-364-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



