Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

D-(-)-Ribose

Glass bottle for solids, 50 g
Catalog # SERA34460.02
Marketsource
Supplier:   Serva
D-(-)-Ribose
Catalog # SERA34460.02
Supplier:  Serva
Marketsource
CAS Number:  50-69-1
D-(-)-Ribose

Glass bottle for solids, 50 g

CAS Number:  50-69-1
Price
Inquire For Price
Log in for availability
1 * 50 g
Product Details & Documents
  • CAS Number:50-69-1
  • Formula:C₅H₁₀O₅
  • MDL Number:MFCD00135453
  • Molecular Weight:150.13 g/mol
  • Merck Index:12,08371
  • Boiling Pt:331 °C (1013 hPa)
  • Melting Pt:90…95 °C
  • Density:0.8 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-059-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications