Polysorbate 80
5 kg
Catalog # SERA33116.02
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
Synonyms: Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate
- CAS Number:9005-65-6
- MDL Number:MFCD00082107
- Melting Pt:100 °C
- Flash Pt:113 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:500-019-9
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:5 kg



