Order Entry
Belgium
Orders LinkContactUsLinkComponent
Polysorbate 80

Polysorbate 80

Glass bottle for solids, 500 g
Catalog # SERA33116.01
Marketsource
Supplier:   Serva
Polysorbate 80
Catalog # SERA33116.01
Supplier:  Serva
Marketsource
CAS Number:  9005-65-6
Polysorbate 80

Glass bottle for solids, 500 g

CAS Number:  9005-65-6
Price
33,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate
  • CAS Number:9005-65-6
  • MDL Number:MFCD00082107
  • Melting Pt:100 °C
  • Flash Pt:113 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:500-019-9

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.