Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Paraformaldehyde

Glass bottle for solids, 100 g
Catalog # SERA31628.01
Flammable
Health
Irritant
Marketsource
Supplier:   Serva
Paraformaldehyde
Catalog # SERA31628.01
Supplier:  Serva
Marketsource
CAS Number:  30525-89-4
Paraformaldehyde

Glass bottle for solids, 100 g

CAS Number:  30525-89-4
Price
20,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Polyoxymethylene
  • CAS Number:30525-89-4
  • Formula:(CH₂O)n
  • MDL Number:MFCD00133991
  • Merck Index:13,07096
  • Boiling Pt:135…136 °C (13 mmHg)
  • Melting Pt:123 °C
  • Density:1.46 g/cm³ (20 °C)
  • Flash Pt:70 °C
  • Storage Temperature:Ambient
  • UN:2213
  • ADR:4.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications