Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Neutral red

Glass bottle for solids, 100 g
Catalog # SERA30305.02
Marketsource
Supplier:   Serva
Neutral red
Catalog # SERA30305.02
Supplier:  Serva
Marketsource
CAS Number:  553-24-2
Neutral red

Glass bottle for solids, 100 g

CAS Number:  553-24-2
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  N2,N2,7-Trimethylphenazine-2,8-diamine hydrochloride, C.I. 50040, 3-Amino-7-dimethyl amino-2-methylphenazine hydrochloride, N8,N8,3-Trimethyl-2,8-phenazinediamine monohydrochloride, Toluylene red, Basic Red 5
  • CAS Number:553-24-2
  • Formula:C₁₅H₁₇ClN₄
  • MDL Number:MFCD00012651
  • Molecular Weight:288.78 g/mol
  • Merck Index:12,06571
  • Melting Pt:290 °C
  • Storage Temperature:Ambient
  • EINECS:209-035-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications