Order Entry
Belgium
Orders LinkContactUsLinkComponent
Neutral red

Neutral red

Glass bottle for solids, 25 g
Catalog # SERA30305.01
Marketsource
Supplier:   Serva
Neutral red
Catalog # SERA30305.01
Supplier:  Serva
Marketsource
CAS Number:  553-24-2
Neutral red

Glass bottle for solids, 25 g

CAS Number:  553-24-2
Price
80,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  N2,N2,7-Trimethylphenazine-2,8-diamine hydrochloride, C.I. 50040, 3-Amino-7-dimethyl amino-2-methylphenazine hydrochloride, N8,N8,3-Trimethyl-2,8-phenazinediamine monohydrochloride, Toluylene red, Basic Red 5
  • CAS Number:553-24-2
  • Formula:C₁₅H₁₇ClN₄
  • MDL Number:MFCD00012651
  • Molecular Weight:288.78 g/mol
  • Merck Index:12,06571
  • Melting Pt:290 °C
  • Storage Temperature:Ambient
  • EINECS:209-035-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.