Sodium molybdate(VI) dihydrate
Glass bottle for solids, 50 g
Catalog # SERA30207.01
Marketsource
Supplier: Serva
Sodium molybdate(VI) dihydrate
Catalog # SERA30207.01
Supplier: Serva
Marketsource
CAS Number: 10102-40-6
Jump to:
Product Details & Documents
Synonyms: Molybdic acid, sodium salt dihydrate, Molybdic acid sodium salt dihydrate, Molybdic acid disodium salt dihydrate, di-Sodium molybdate dihydrate, Sodium molybdate dihydrate, Sodium molybdenum oxide dihydrate
- CAS Number:10102-40-6
- Formula:Na₂MoO₄·2H₂O
- MDL Number:MFCD00149170
- Molecular Weight:241.95 g/mol
- Merck Index:13,08718
- Melting Pt:687 °C
- Density:3.28 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-551-7
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



