Bisacrylamide
50 g
Catalog # SERA29196.02
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
Synonyms: Bis (N,N'-methylenebisacrylamide), N,N-Methylenebisacrylamide, N,N'-Methylenediacrylamide, N,N’-Methylenebisacrylamide, N,N'-Methylenebisacrylamide, BIS
- CAS Number:110-26-9
- Formula:(H₂C=CHCONH)₂CH₂
- MDL Number:MFCD00008625
- Molecular Weight:154.17 g/mol
- Boiling Pt:125 °C (3.33 hPa)
- Melting Pt:185 °C
- Density:1.235 g/cm³ (20 °C)
- Flash Pt:138 °C
- Storage Temperature:Refrigerator
- EINECS:203-750-9
Specifications
Specifications
- Pk:50 g



