Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

D-(+)-Mannose

Glass bottle for solids, 10 g
Catalog # SERA28460.01
Marketsource
Supplier:   Serva
D-(+)-Mannose
Catalog # SERA28460.01
Supplier:  Serva
Marketsource
CAS Number:  3458-28-4
D-(+)-Mannose

Glass bottle for solids, 10 g

CAS Number:  3458-28-4
Price
Inquire For Price
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
  • CAS Number:3458-28-4
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00064122
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,05772
  • Boiling Pt:376 °C (1013 hPa)
  • Melting Pt:133 °C
  • Density:1.539 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:222-392-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications