Lanthanum(III) nitrate
50 g
Catalog # SERA27507.04
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
- CAS Number:10099-59-9
- Formula:LaN₃O₉
- MDL Number:MFCD00011067
- Molecular Weight:324.92
- Storage Temperature:Refrigerator
- EINECS:233-238-0
- UN:1477
- ADR:5.1,II
Specifications
Specifications
- Pk:50 g



