Order Entry
Belgium
Orders LinkContactUsLinkComponent
Hypoxanthine

Hypoxanthine

Glass bottle for solids, 25 g
Catalog # SERA26040.02
Marketsource
Supplier:   Serva
Hypoxanthine
Catalog # SERA26040.02
Supplier:  Serva
Marketsource
CAS Number:  68-94-0
Hypoxanthine

Glass bottle for solids, 25 g

CAS Number:  68-94-0
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  1H-Purin-6-one, 1,9-Dihydro-6H-purin-6-one, Purin-6(1H)-one, 6-Purinone
  • CAS Number:68-94-0
  • Formula:C₅H₄N₄O
  • MDL Number:MFCD00005725
  • Molecular Weight:136.11 g/mol
  • Merck Index:13,04895
  • Melting Pt:> 310 °C
  • EINECS:200-697-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.