Order Entry
Belgium
Orders LinkContactUsLinkComponent
Standard reference materials and buffer solutions, CertiPUR®, Supelco®

Standard reference materials and buffer solutions, CertiPUR®, Supelco®

Potassium tetraoxalate dihydrate, 1,681 (25°C), Glass bottle for solids, 1×25 g, 25 g
Catalog # 1.01961.0025
Supplier:   Merck
Standard reference materials and buffer solutions, CertiPUR®, Supelco®
Catalog # 1.01961.0025
Supplier:  Merck
CAS Number:  
Standard reference materials and buffer solutions, CertiPUR®, Supelco®

Potassium tetraoxalate dihydrate, 1,681 (25°C), Glass bottle for solids, 1×25 g, 25 g

CAS Number:  
Price
204,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Materials for the precise calibration and monitoring of pH instruments.

  • Traceable to NIST primary standard and PTB
  • Maximum deviation ±0,003 pH units
  • Measured in an independent accredited calibration laboratory for the pH value measurement (DKD-K-14301)
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Description:
    Potassium tetraoxalate dihydrate
  • Pk Info:
    1×25 g
  • pH value:
    1,681 (25°C)
  • Cat. No.:
    1.01961.0025
  • M-Clarity quality level:
    MQ100

Specifications