D-(+)-Glucose
5 kg
Catalog # SERA22700.03
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose
- CAS Number:50-99-7
- Formula:C₆H₁₂O₆
- MDL Number:MFCD00148912
- Molecular Weight:180.16 g/mol
- Merck Index:14,04459
- Melting Pt:146 °C
- Density:1.54 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-075-1
Specifications
Specifications
- Pk:5 kg



