D-(+)-Glucose
Glass bottle for solids, 100 g
SERA22700.01
Marketsource
: Serva
: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose
- :50-99-7
- :C₆H₁₂O₆
- :MFCD00148912
- :180.16 g/mol
- :14,04459
- :146 °C
- :1.54 g/cm³ (20 °C)
- :Ambient
- :200-075-1
- Pack type:Glass bottle for solids
- Pk:100 g