Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Forskolin

Glass bottle for solids, 5 mg
Catalog # SERA21780.02
Marketsource
Supplier:   Serva
Forskolin
Catalog # SERA21780.02
Supplier:  Serva
Marketsource
CAS Number:  66575-29-9
Forskolin

Glass bottle for solids, 5 mg

CAS Number:  66575-29-9
Price
Inquire For Price
Log in for availability
1 * 5 mg
Product Details & Documents
Synonyms:  (3R,4aR,5S,6S,6aS,10S,10aR,10bS)-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-3-vinyldodecahydro-1H-benzo[f]chromen-5-yl acetate
  • CAS Number:66575-29-9
  • Formula:C₂₂H₃₄O₇
  • MDL Number:MFCD00082317
  • Molecular Weight:410.51 g/mol
  • EINECS:266-410-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg

Specifications