Fast green FCF
Glass bottle for solids, 5 g
Catalog # SERA21295.01
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
Synonyms: Light green, Sodium 2-((4-(ethyl(3-sulphonatobenzyl)amino)phenyl)(4-(ethyl(3-sulphonatobenzyl)iminio)cyclohexa-2,5-dien-1-ylidene)methyl)-5-hydroxybenzenesulphonate, CI 42053, Dihydrogen (ethyl)[4-[4-[ethyl(3-sulphonatobenzyl)amino](4-hydroxy-2-sulphonatobenzhydrylidene]cyclohexa-2,5-dien-1-ylidene](3-sulphonatobenzyl)ammonium, disodium salt
- CAS Number:2353-45-9
- Formula:C₃₇H₃₄N₂Na₂O₁₀S₃
- MDL Number:MFCD00013053
- Molecular Weight:808.86 g/mol
- Merck Index:13,03970
- Melting Pt:290 °C
- Storage Temperature:Ambient
- EINECS:219-091-5
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



