Dextran, M.W. ~ 200,000
Glass bottle for solids, 500 g
Catalog # SERA18694.02
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
- CAS Number:9004-54-0
- Formula:(C₆H₁₀O₅)n
- MDL Number:MFCD00130935
- EINECS:232-677-5
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



