Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Sodium deoxycholate

Glass bottle for solids, 25 g
Catalog # SERA18330.02
Irritant
Marketsource
Supplier:   Serva
Sodium deoxycholate
Catalog # SERA18330.02
Supplier:  Serva
Marketsource
CAS Number:  302-95-4
Sodium deoxycholate

Glass bottle for solids, 25 g

CAS Number:  302-95-4
Price
39,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Deoxycholic acid sodium salt, 3α,12α-Dihydroxy-5β-cholan-24-oic acid sodium salt, Sodium 3α,12α-dihydroxy-5β-cholanoate
  • CAS Number:302-95-4
  • Formula:C₂₄H₃₉NaO₄
  • MDL Number:MFCD00064139
  • Molecular Weight:414.56 g/mol
  • Melting Pt:357…365 °C
  • Storage Temperature:Ambient
  • EINECS:206-132-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.