Celite® 545, filter aid
25 kg
Catalog # SERA16391.04
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
Synonyms: DE, Diatomaceous earth, flux-calcined
- CAS Number:68855-54-9
- Formula:SiO₂
- MDL Number:MFCD00132803
- Molecular Weight:60.08 g/mol
- EINECS:272-489-0
Specifications
Specifications
- Pk:25 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



