Orders LinkContactUsLinkComponent
L(+)-Ascorbic acid
Glass bottle for solids, 500 g
  SERA14030.03
Marketsource
 :   Serva
L(+)-Ascorbic acid
  SERA14030.03
 :  Serva
Marketsource
 :  50-81-7
L(+)-Ascorbic acid
L(+)-Ascorbic acid
Glass bottle for solids, 500 g
 :  50-81-7
1 * 500 g
 :  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  •  :50-81-7
  •  :C₆H₈O₆
  •  :MFCD00064328
  •  :176.13 g/mol
  •  :13,00837
  •  :553 °C (1013 hPa)
  •  :190…192 °C
  •  :1.694 g/cm³ (20 °C)
  •  :Ambient
  •  :200-066-2
  •  :exempt
  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g