Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Agarose

Glass bottle for solids, 100 g
Catalog # SERA11397.03
Marketsource
Supplier:   Serva
Agarose
Catalog # SERA11397.03
Supplier:  Serva
Marketsource
CAS Number:  9012-36-6
Agarose
Agarose

Glass bottle for solids, 100 g

CAS Number:  9012-36-6
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:9012-36-6
  • MDL Number:MFCD00081294
  • Melting Pt:60…90 °C
  • Storage Temperature:Ambient
  • EINECS:232-731-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.