L(-)-Malic acid
Glass bottle for solids, 100 g
Catalog # SERA11050.03
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
Synonyms: L(-)-Malic acid, (S)-(-)-Hydroxy-1,4-butanedioic acid, (S)-(-)-Hydroxysuccinic acid, (S)-2-hydroxysuccinic acid, L(-)-Hydroxysuccinic acid, L(-)-Hydroxy-1,4-butanedioic acid, L-(–)-Malic Acid
- CAS Number:97-67-6
- Formula:C₄H₆O₅
- MDL Number:MFCD00064213
- Molecular Weight:134.09 g/mol
- Merck Index:13,05730
- Boiling Pt:322 °C (1013 hPa)
- Melting Pt:101…104 °C
- Density:1.595 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:202-601-5
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



