Order Entry
Belgium
Orders LinkContactUsLinkComponent
L(-)-Malic acid

L(-)-Malic acid

Glass bottle for solids, 25 g
Catalog # SERA11050.02
Irritant
Marketsource
Supplier:   Serva
L(-)-Malic acid
Catalog # SERA11050.02
Supplier:  Serva
Marketsource
CAS Number:  97-67-6
L(-)-Malic acid

Glass bottle for solids, 25 g

CAS Number:  97-67-6
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  L(-)-Malic acid, (S)-(-)-Hydroxy-1,4-butanedioic acid, (S)-(-)-Hydroxysuccinic acid, (S)-2-hydroxysuccinic acid, L(-)-Hydroxysuccinic acid, L(-)-Hydroxy-1,4-butanedioic acid, L-(–)-Malic Acid
  • CAS Number:97-67-6
  • Formula:C₄H₆O₅
  • MDL Number:MFCD00064213
  • Molecular Weight:134.09 g/mol
  • Merck Index:13,05730
  • Boiling Pt:322 °C (1013 hPa)
  • Melting Pt:101…104 °C
  • Density:1.595 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:202-601-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.