Orders LinkContactUsLinkComponent
Dropping bottles, PP, Nalgene®
125 ml
  216-4432
   Low Manufacturing Impact
Dropping bottles, PP, Nalgene®
  216-4432
   Low Manufacturing Impact
Dropping bottles, PP, Nalgene®
Dropping bottles, PP, Nalgene®
125 ml
71,90 €
1 * 6 ST
PPCO with non-drip closure and cap made from blue PP.
  • Autoclavable even when full with tips uncapped
  • Leakproof closure
  • Integral non-drip closure with attached cap
Ideal for sterile solutions.
  • Capacity:
    125 ml
  • Cat. No.:
    216-4432
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact