Orders LinkContactUsLinkComponent
Nalgene™ Screw Caps
Nalgene™ Screw Caps
Replacement closure, 53B, White, PP
Catalog # NALG71-2151-0053
   Low Manufacturing Impact
Nalgene™ Screw Caps
Catalog # NALG71-2151-0053
   Low Manufacturing Impact
Nalgene™ Screw Caps
Nalgene™ Screw Caps
Replacement closure, 53B, White, PP
Price
93,80 €
Log in for availability
1 * 12 ST
Product Details & Documents
PP, coloured.
  • Very good chemical resistance
  • Heat resistant up to 135 °C
  • Autoclavable
Screw caps without inserts for Nalgene® narrow and wide mouth bottles, as well as Erlenmeyer flasks with thread.
Specifications
  • Cap Size:
    53B
  • Colour:
    White
  • Description:
    Replacement closure
  • Material:
    PP
  • Seq:
    37
  • Cat. No.:
    NALG71-2151-0053
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact