Nalgene™ Screw Caps
Replacement closure, 53 mm, —, PP
Catalog # NALG71-2240-1053
Supplier: Thermo Fisher Scientific
Nalgene™ Screw Caps
Catalog # NALG71-2240-1053
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
PP, coloured.
- Very good chemical resistance
- Heat resistant up to 135 °C
- Autoclavable
Screw caps without inserts for Nalgene® narrow and wide mouth bottles, as well as Erlenmeyer flasks with thread.
Specifications
- Cap Size:53 mm
- Colour:—
- Description:Replacement closure
- Material:PP
- Seq:50
- Cat. No.:NALG71-2240-1053
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



