Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer solutions

Buffer solutions

Type L4697, 7,00, ±0,010, 25 °C, 60 Ampoul
Catalog # SCOI285138221
Supplier:   SI Analytics
Buffer solutions
Catalog # SCOI285138221
Supplier:  SI Analytics
CAS Number:  
Buffer solutions
Buffer solutions

Type L4697, 7,00, ±0,010, 25 °C, 60 Ampoul

CAS Number:  
Price
224,00 €
Log in for availability
1 * 60 Ampul
Product Details & Documents

Buffer solutions intended for extremely precise calibration.

  • Highest measurement reliability
  • No preservative agents
  • Hermetically sealed in the glass ampoule
  • Sterilised with hot steam

Certifications: According to DIN 19 266.

Specifications

Specifications

  • Pk:
    60 Ampoul
  • Reference temperature:
    25 °C
  • Description:
    Type L4697
  • pH accuracy:
    ±0,010
  • pH value:
    7,00

Specifications