Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Micro filter funnels, DURAN®

10 mm, 2, 2 ml, 6 mm
  SCOT258575207
Micro filter funnels, DURAN®
  SCOT258575207
Micro filter funnels, DURAN®
Micro filter funnels, DURAN®

10 mm, 2, 2 ml, 6 mm

 
46,00 €
   
1 * 1 ST
 

DURAN®, borosilicate glass 3.3, clear.

  • For filtration of very small volumes
  • Highly resistant to most chemicals such as solvents, acids and alkalis
  • Very good temperature resistance

With integrated sintered glass filter plate.

 
  • Plate Ø:
    10 mm
  • Porosity:
    2
  • Capacity:
    2 ml
  • Stem O-Ø:
    6 mm