VWR®, Calibration Weights, Class M2
10 g, 6 ± mg
Catalog # 611-8373
Supplier: VWR Collection
Jump to:
Product Details & Documents
Test weights in either stainless steel or aluminium.
Delivery information: Supplied in a plastic box.
Documents
Specifications
- Weight:10 g
- Seq:07
- Tolerance:6 ± mg