VWR®, Calibration Weights, Class M2
500 g, 75 ± mg
Catalog # 611-8362
Supplier: VWR Collection
Jump to:
Product Details & Documents
Test weights in either stainless steel or aluminium.
Delivery information: Supplied in a plastic box.
Documents
Specifications
- Weight:500 g
- Seq:28
- Tolerance:75 ± mg