Orders LinkContactUsLinkComponent
VWR®, Calibration Weights, Class M2
100 g, 16  ± mg
Catalog # 611-8359
Supplier:   VWR Collection
VWR®, Calibration Weights, Class M2
Catalog # 611-8359
Supplier:  VWR Collection
VWR®, Calibration Weights, Class M2
VWR®, Calibration Weights, Class M2
100 g, 16  ± mg
Price
29,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
Test weights in either stainless steel or aluminium.
Delivery information: Supplied in a plastic box.
Specifications
  • Weight:
    100 g
  • Seq:
    26
  • Tolerance:
    16  ± mg