Order Entry
Belgium
Orders LinkContactUsLinkComponent
Separating funnels, Squibb, Nalgene®

Separating funnels, Squibb, Nalgene®

1000 ml, 53 mm
Catalog # 527-2104
   Low Manufacturing Impact
Separating funnels, Squibb, Nalgene®
Catalog # 527-2104
   Low Manufacturing Impact
Separating funnels, Squibb, Nalgene®
Separating funnels, Squibb, Nalgene®

1000 ml, 53 mm

Price
413,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

FEP translucent, TFE stopcock, ETFE screw closure.

  • Leakproof closure and stopcock
  • Autoclavable except for stopcock assembly
  • TFE stopcock requires no lubrication

Robust, transparent, non stick for easy cleaning, non wetting for complete draining and impervious to any chemical used in a separating funnel.

Documents

Specifications
  • Capacity:
    1000 ml
  • Ø cap:
    53 mm
  • Cat. No.:
    527-2104
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Replacement stopcock assembly
    Replacement stopcock assembly
    NALG71-4301-1050
    Log in for availability
    1 * 1 ST - 147,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.