Order Entry
Belgium
Orders LinkContactUsLinkComponent
Separating funnels, Squibb, Nalgene®

Separating funnels, Squibb, Nalgene®

250 ml, 33 mm
Catalog # 527-2102
   Low Manufacturing Impact
Separating funnels, Squibb, Nalgene®
Catalog # 527-2102
   Low Manufacturing Impact
Separating funnels, Squibb, Nalgene®
Separating funnels, Squibb, Nalgene®

250 ml, 33 mm

Price
240,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

FEP translucent, TFE stopcock, ETFE screw closure.

  • Leakproof closure and stopcock
  • Autoclavable except for stopcock assembly
  • TFE stopcock requires no lubrication

Robust, transparent, non stick for easy cleaning, non wetting for complete draining and impervious to any chemical used in a separating funnel.

Documents

Specifications
  • Capacity:
    250 ml
  • Ø cap:
    33 mm
  • Cat. No.:
    527-2102
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Replacement stopcock assembly
    Replacement stopcock assembly
    NALG71-4301-1050
    Log in for availability
    1 * 1 ST - 147,00 €