Separating funnels, Squibb, Nalgene®
125 ml, 28 mm
Catalog # 527-2101
Supplier: Thermo Fisher Scientific
Separating funnels, Squibb, Nalgene®
Catalog # 527-2101
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
FEP translucent, TFE stopcock, ETFE screw closure.
- Leakproof closure and stopcock
- Autoclavable except for stopcock assembly
- TFE stopcock requires no lubrication
Robust, transparent, non stick for easy cleaning, non wetting for complete draining and impervious to any chemical used in a separating funnel.
Documents
NALG4301-0250_TDS_EN
Datasheet
Specifications
- Capacity:125 ml
- Ø cap:28 mm
- Cat. No.:527-2101
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact
Accessories
Product Information
- Catalog #AvailabilityPrice
- Replacement stopcock assembly
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



