Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR®, Calibration Weights, Class M1

VWR®, Calibration Weights, Class M1

2 kg, 100 ± mg
Catalog # 611-8269
Supplier:   VWR Collection
VWR®, Calibration Weights, Class M1
Catalog # 611-8269
Supplier:  VWR Collection
VWR®, Calibration Weights, Class M1
VWR®, Calibration Weights, Class M1

2 kg, 100 ± mg

Price
207,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.

Delivery information: All weights are supplied in plastic cases.

Specifications
  • Weight:
    2 kg
  • Seq:
    45
  • Tolerance:
    100 ± mg
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.