Order Entry
Belgium
Orders LinkContactUsLinkComponent
Dropping bottles, LDPE, Nalgene®

Dropping bottles, LDPE, Nalgene®

15 ml, LDPE, translucent
Catalog # 215-7172
   Low Manufacturing Impact
Dropping bottles, LDPE, Nalgene®
Catalog # 215-7172
   Low Manufacturing Impact
Dropping bottles, LDPE, Nalgene®
Dropping bottles, LDPE, Nalgene®

15 ml, LDPE, translucent

Price
63,80 €
Log in for availability
1 * 25 ST
Product Details & Documents

LDPE, translucent or LDPE, white with white PP screw cap and LDPE dosing tip.

  • Good chemical resistance, heat resistant up to +80 °C
  • Drop-control tip snaps securely into place, drops are dispensed individually
  • Flexible for easy handling, leakproof
  • Delivers 40 µl drops (based on water; viscosity affects drop size)

These dropping bottles provide reliable, repeatable dispensing of reagents and other liquids. They serve as a very good alternative to pipettes or other conventional dispensing devices. They are suitable for most biotech, diagnostic and pharmaceutical applications. White material, ideal for UV light-sensitive products.

Specifications
  • Capacity:
    15 ml
  • Material:
    LDPE, translucent
  • Seq:
    05
  • Cat. No.:
    215-7172
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.