Order Entry
Belgium
Orders LinkContactUsLinkComponent
Dropping bottles, LDPE, Nalgene®

Dropping bottles, LDPE, Nalgene®

8 ml, LDPE, translucent
Catalog # 215-7171
   Low Manufacturing Impact
Dropping bottles, LDPE, Nalgene®
Catalog # 215-7171
   Low Manufacturing Impact
Dropping bottles, LDPE, Nalgene®
Dropping bottles, LDPE, Nalgene®

8 ml, LDPE, translucent

Price
60,80 €
Log in for availability
1 * 25 ST
Product Details & Documents

LDPE, translucent or LDPE, white with white PP screw cap and LDPE dosing tip.

  • Good chemical resistance, heat resistant up to +80 °C
  • Drop-control tip snaps securely into place, drops are dispensed individually
  • Flexible for easy handling, leakproof
  • Delivers 40 µl drops (based on water; viscosity affects drop size)

These dropping bottles provide reliable, repeatable dispensing of reagents and other liquids. They serve as a very good alternative to pipettes or other conventional dispensing devices. They are suitable for most biotech, diagnostic and pharmaceutical applications. White material, ideal for UV light-sensitive products.

Specifications
  • Capacity:
    8 ml
  • Material:
    LDPE, translucent
  • Seq:
    03
  • Cat. No.:
    215-7171
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact