Orders LinkContactUsLinkComponent
Separating funnels, spherical, DURAN®
Separating funnels, spherical, DURAN®
500 ml
Catalog # 527-1412
Supplier:   DWK Life Sciences
Separating funnels, spherical, DURAN®
Catalog # 527-1412
Supplier:  DWK Life Sciences
Separating funnels, spherical, DURAN®
Separating funnels, spherical, DURAN®
500 ml
Price
232,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN® borosilicate glass 3.3, clear, stopcock with glass key and plastic stopper.
  • Without graduation
  • Very good chemical resistance
  • Very good temperature resistance
Specifications
  • Capacity:
    500 ml